Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIN1 Mouse anti-Human, Mouse, Clone:3B6F10, Abnova™

Catalog No. 89333570 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-570 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-570 Supplier Abnova Corporation Supplier No. MAB17313
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human BIN1.

This gene encodes several isoforms of a nucleocytoplasmic adaptor protein, one of which was initially identified as a MYC-interacting protein with features of a tumor suppressor. Isoforms that are expressed in the central nervous system may be involved in synaptic vesicle endocytosis and may interact with dynanim, synaptojanin, endophilin, and clathrin. Isoforms that are expressed in muscle and ubiquitously expressed isoforms localize to the cytoplasm and nucleus and activate a caspase-independent apoptotic process. Studies in mouse suggest that this gene plays an important role in cardiac muscle development. Alternate splicing of the gene results in ten transcript variants encoding different isoforms. Aberrant splice variants expressed in tumor cell lines have also been described. [provided by RefSeq]

Specifications

Antigen BIN1
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 3B6F10
Conjugate Unconjugated
Description bridging integrator 1
Dilution ELISA (1:10000)
Flow Cytometry (1:200-400)
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1000)
Western Blot (1:500-1:2000)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene BIN1
Gene Alias AMPH2, AMPHL, DKFZp547F068, MGC10367, SH3P9
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 189-398 of of human BIN1 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 274
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.