Learn More
Invitrogen™ BIN1 Polyclonal Antibody

Description
BIN1 Polyclonal Antibody detects endogenous levels of BIN1 protein.
Specifications
Specifications
| Antigen | BIN1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4 |
| Gene | BIN1 |
| Gene Accession No. | O00499, O08539, O08839 |
| Gene Alias | ALP-1; AMPH 2; Amph2; amphiphysin 2; amphiphysin II; amphiphysin IIamph2; amphiphysin-like; amphiphysin-like protein; AMPHL; Bin1; box dependant MYC interacting protein 1; box-dependent myc-interacting protein 1; BRAMP-2; bridging integrator 1; DKFZp547F068; MGC10367; MGC105358; myc box dependent interacting protein 1; myc box-dependent-interacting protein 1; SH3 domain-containing protein 9; SH3P9; similar to bridging integrator 1 isoform 9 |
| Gene Symbols | BIN1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.