Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BIN2 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297015DL594
Description
BIN2 Polyclonal specifically detects BIN2 in Human samples. It is validated for Western Blot, ELISA.Specifications
BIN2 | |
Polyclonal | |
Western Blot, ELISA | |
BRAP1, BRAP-1, breast cancer associated protein BRAP1, Breast cancer-associated protein 1, bridging integrator 2, bridging integrator-2 | |
Produced in rabbits immunized with purified, recombinant BIN2 (Q9UBW5-1; Met1-Asn244). | |
0.1 mL | |
Cell Cycle and Replication | |
51411 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction