Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BIN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558486
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558486 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558486 Supplier Invitrogen™ Supplier No. PA558486
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EKPVRTPEAK ENENIHNQNP EELCTSPTLM TSQVASEPGE AKKMEDKEKD NKLISADSSE GQDQLQVSMV PENNNLTAPE PQEEVSTSEN PQL Highest antigen sequence identity to the following orthologs: Mouse - 29%, Rat - 31%.

BAR proteins, which are characterized by a common N-terminal BAR domain, are a family of adaptor proteins implicated in a diverse set of cellular processes. BIN1 (bridging integrator 1) and BIN2 are BAR proteins. The human BIN2 gene is located at chromosome 4q22.1, and encodes a 565-amino acid protein with a predicted molecular weight of ~62 kDa. It shows preferential expression in hematopoietic tissues, with highest mRNA levels in spleen and peripheral blood leukocytes and also high levels in thymus, colon, and placenta. BIN2 has been shown to migrate with an apparent mobility of ~80 kDa, and interact with BIN1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BIN2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BIN2
Gene Accession No. Q9UBW5
Gene Alias BIN2; BRAP1; BRAP-1; breast cancer associated protein BRAP1; breast cancer-associated protein 1; Bridging integrator 2; bridging integrator-2
Gene Symbols BIN2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BIN2. Recombinant protein control fragment (Product #RP-97670).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51411
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.