Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BIN3 Antibody (OTI8E9), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272051PEATT594
Description
BIN3 Monoclonal specifically detects BIN3 in Human, Mouse, Rat, Canine, Monkey, Primate samples. It is validated for Flow Cytometry.Specifications
BIN3 | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
BIN3 | |
Full length recombinant protein of human BIN3 (NP_061158) produced in HEK293T cells. | |
0.1 mL | |
55909.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
OTI8E9 | |
Flow Cytometry | |
bridging integrator 3, MGC14978 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Canine, Monkey, Primate | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction