Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BIN3 Antibody (OTI8E9), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272051PCP
Description
BIN3 Monoclonal specifically detects BIN3 in Human, Mouse, Rat, Canine, Monkey, Primate samples. It is validated for Flow Cytometry.Specifications
| BIN3 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| BIN3 | |
| Full length recombinant protein of human BIN3 (NP_061158) produced in HEK293T cells. | |
| 0.1 mL | |
| 55909.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI8E9 | |
| Flow Cytometry | |
| bridging integrator 3, MGC14978 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey, Primate | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction