Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BIN3 Monoclonal Antibody (OTI5E3)
GREENER_CHOICE

Catalog No. PIMA525302
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525302 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525302 Supplier Invitrogen™ Supplier No. MA525302
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BIN3 Monoclonal Antibody for Flow, ICC/IF, IHC (P), Western Blot

Involved in cytokinesis and septation where it has a role in the localization of F-actin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BIN3
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI5E3
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 1% BSA and 0.02% sodium azide; pH 7.3
Gene BIN3
Gene Accession No. Q68FW8, Q9NQY0
Gene Alias 1700015F07Rik; Bin3; Bridging integrator 3; bridging integrator-3
Gene Symbols BIN3
Host Species Mouse
Immunogen Full length human recombinant protein of BIN3 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361065, 55909, 608969
Target Species Human, Rat, Canine, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.