Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BIN3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554882
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554882 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554882 Supplier Invitrogen™ Supplier No. PA554882
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PKTVERDFER EYGKLQQLEE QTRRLQKDMK KSTDADLAMS KSAVKISLDL LSNPLCEQDQ D Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 97%.

Involved in cytokinesis and septation where it has a role in the localization of F-actin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BIN3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BIN3
Gene Accession No. Q9NQY0
Gene Alias 1700015F07Rik; Bin3; Bridging integrator 3; bridging integrator-3
Gene Symbols BIN3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BIN3. Recombinant protein control fragment (Product #RP-93802).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55909
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.