Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BIN3 Monoclonal Antibody (OTI8E9)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA525348
Description
BIN3 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Involved in cytokinesis and septation where it has a role in the localization of F-actin.
Specifications
BIN3 | |
Monoclonal | |
0.69 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NQY0 | |
BIN3 | |
Full length human recombinant protein of BIN3 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human, Canine, Monkey | |
Antibody | |
IgG2a |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
OTI8E9 | |
Unconjugated | |
BIN3 | |
1700015F07Rik; Bin3; Bridging integrator 3; bridging integrator-3 | |
Mouse | |
Affinity chromatography | |
RUO | |
55909, 608969 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction