Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ BINA
GREENER_CHOICESDP

Catalog No. 404850 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-485-0 50 mg
40-481-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 40-485-0 Supplier R&D Systems™ Supplier No. 4048/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective positive allosteric modulator of mGlu2

Selective positive allosteric modulator of mGlu2 (EC50 = 33.2 nM in CHO cells expressing human mGlu2). Displays no effect on glutamate-induced activation of other mGlu receptor subtypes. Exhibits antipsychotic and anxiolytic properties in mice.

Chemical Identifiers

CAS 866823-73-6
Molecular Formula C30H30O4

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 454.56
Purity >98%
Molecular Weight (g/mol) 454.566
Chemical Name or Material BINA
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.