Learn More
Description
- It is used to extract genomic DNA from plants, animals, bacteria, yeast, algae, and fungi, soil and environmental samples.
- Binding Matrix is a component included in the FastDNA and all FastDNA Spin Kits and is used to bind DNA from a lysate.
- Once bound, impurities and inhibitors are washed away with the SEWS-M solution.
- The resulting pure DNA is then eluted from the Binding Matrix with either an elution solution or DDI water.
Specifications
Specifications
| Sample Type | Seeds, Spores, Bacteria (Gram +/-), Animal Tissues, Feces, Food, Fungi, Plants, Roots, Soil, Wastewater, Yeast |
| For Use With (Equipment) | FastPrep-24, FastPrep 120, FastPrep-96, SuperFastPrep-1 |
| For Use With (Application) | Sample Preparation, Sample Lysis, Nucleic Acid Isolation and Purification, Molecular Biology, Gene Expression, Microbiological Analysis, Gene Function, Gene Regulation |
| Product Type | Binding Matrix |
| Quantity | 2 x 30 mL |
| Sample Volume | Up to 500 mg Per Prep |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.