Learn More
National Optical Binocular Cordless LED Microscope
SCI_ED
Catalog No. s81776 Shop All Motic Swift Line Products
Click to view available options
Objective Types:
Achromatic
Catalog No. S81776
Supplier: National Optical 215RLED
Only null left
Add to Cart
Add to Cart
A new standard of precision optics, highly engineered mechanics and very rugged construction, all at a little more cost than a budget microscope.
Advanced Biological Binocular microscope with precision optics. For use by students or professionals. Model 215 has a Seidentopf head for comfortable viewing, coaxial focusing, low drive mechanical stage and LED illumination with conveniently located intensity control. LED light rated up to 50,000 hours.
OPTICAL SYSTEM
  • Widefield 10x eyepiece with pointer. Accepts optional #965-160 eyepiece reticle.
  • 30° inclined binocular viewing head rotates 360° for easy sharing by more than one student.
  • DIN standard 4x (0.10 N.A.), 10x (0.25 N.A.), 40xR (0.65 N.A.) and 100xR (1.25 N.A.) objectives are supplied for 40x, 100x, 400x and 1000x magnification. All are achromatic, parfocalled, parcentered and color-coded.
  • 4-hole objective turret, reverse position, ball-bearing mounted for smooth, precise changes in magnification.
ILLUMINATION
  • Cool running LED light with intensity control rated up to 50,000 hours of life.
  • Rechargeable batteries can be recharged up to 1000 times.
  • 8 hour charge provides up to 50 hours of use.
  • Recharge adapter shuts off when batteries are fully charged to prevent overcharging and permits conventional light operation by plugging into an electrical outlet.
FOCUSING
  • Coaxial coarse and fine focusing controls.
  • Slip clutch prevents damage to focusing system.
  • Tension adjustment eliminates stage drift.
  • Safety rack stop prevents damage to slides and objective lenses.
STAGE
  • Large 120 mm x 120 mm stage, with professional quality, low profile mechanical specimen holder with low-position coaxial x-y controls.
  • 1.25 N.A. Abbe condenser with spiral focusing mount, iris diaphragm, built-in frosted filter.
Specifications
Eyepiece WF10X/18 mm
Head Style Binocular
Lighting Type LED
Magnification Power 4X, 10X, 40XR, 100XR
Model 215-RLED
Objective Types Achromatic
Product Type Binocular LED Microscope
Includes Dustcover
Material Metal, Aluminum
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.