Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Bio-Bag™ Environmental Chambers

Disposable systems for cultivation and isolation of specific bacteria and cultures

Supplier:  BD 261212

 View more versions of this product

Catalog No. 14-910-55


Requires :

Media and plates


This item is not returnable. View return policy

Description

Description

The Bio-Bag™ System incorporates traditional gas-generation methods into a reliable pouch system, eliminating the need to add water.

  • Create anaerobic, CO2-enriched aerobic, or microaerophilic environments, depending on bag type
  • Transparent pouches allow observation without disturbing the environment

Bio-Bag Type A (Anaerobic)

  • Maintains anaerobic conditions for 96 hours
  • 100-Pack holds one 100 x 15mm Petri dish or MIC microdilution plate
  • 25-Pack is ideal for smaller workloads
  • Multi-Plate version holds three 100 x 15mm multi-plate dishes or two API ID strips

Bio-Bag Type C (CO2-enriched aerobic)

  • Maintains CO2-enriched aerobic environment for 48 hours
  • Holds one 100 x 15mm Petri dish or MIC plate
  • Gas generators (14-910-33, also sold separately) have a two-year shelf life

Bio-Bag Type Cfj (Microaerophilic)

  • For microaerophilic environment
  • Supports growth and isolation of Campylobacter jejuni
  • 100-Pack holds one or two 100 x 15mm Petri dishes or MIC plates
  • 25-Pack is designed for smaller workloads
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Environmental Chamber, Type Cfj
Single-use
Petri Dishes (1 or 2)
Bio-Bag
15°C to 30°C
Isolation of Campylobacter jejuni
25 microaerophilic generators
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.