Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio Plas™ Uni-Flex™ Safety Caps

Catalog No. 13879103
Change view
Click to view available options
Color:
Gray
Green
Lavender
Natural
Red
Yellow
For Use With (Equipment):
12 mm Culture or 13 mm Blood Collection Tubes
13 mm Culture Tubes
13 mm Culture and Blood Collection Tubes
16 mm Culture and Blood Collection Tubes
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color For Use With (Equipment)
13-879-103 Yellow 13 mm Culture Tubes
15-182-224 Green 13 mm Culture Tubes
02-706-19 Gray 13 mm Culture Tubes
02-706-34 Red 16 mm Culture and Blood Collection Tubes
15-182-220 Natural 12 mm Culture or 13 mm Blood Collection Tubes
15-182-222 Gray 12 mm Culture or 13 mm Blood Collection Tubes
13-879-099 Lavender 12 mm Culture or 13 mm Blood Collection Tubes
13-879-101 Red 13 mm Culture and Blood Collection Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 13-879-103 Supplier Bio Plas™ Supplier No. 6630
Add to Cart
Add to Cart
This item is not returnable. View return policy

Protects laboratory staff from infectious aerosols

  • Liquid-tight seal ensures sample integrity and prevent spillage of serum samples
  • Designed to hold under centrifugation, agitation, refrigeration and freezing
  • Stretchable, unique, over-locking design to prevent evaporation
  • One-hand application and removal
  • Trace-metal free material

Specifications

Includes Liquid-tight seal
Product Type Safety Cap
Material Polyethylene
Color Yellow
For Use With (Equipment) 13 mm Culture Tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.