Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Current Technologies Bio-Screen™ Biohazard Wipes, Liners, and Rolls
Click to view available options
Dimensions (L x W):
100 ft. x 16 in.
16 x 11 in. (406.4 x 279.4mm)
16 x 16 in. (406.4 x 406.4mm)
3 x 3 in. (76.2 x 76.2mm)
4 x 4 in. (101.6 x 101.6mm)
9 x 8 in. (228.6 x 203.2mm)
Absorbancy:
High
Standard
Super
Description
- White absorbent side of this wipe / liner is low-linting and more absorbent than gauze or paper wipes
- Orange plastic backing of wipe / liner blocks soak through of blood, body fluids, specimens and chemicals (xylene, ketones, formalin) to gloves and surfaces
- Used for cleaning up small / large spills, to wipe pipets, probes, slide prep devices, and to pop caps from blood and specimen tubes
- Limit cross-contamination from gloves as employees touch phones, keyboards, doors, etc.
- Larger sizes can be used as mats and liners. Rolls of material can be cut to desired sizes
- Three thicknesses available: Standard (absorbs 9 times its weight), Heavy (absorbs 12 times its weight), Super (absorbs 25 times its weight)
Specifications
Specifications
Product Type | Biohazard Wipe |
Absorbancy | High |
Length (English) | 3 in. |
Length (Metric) | 76.2 mm |
Width (English) | 3 in. |
Width (Metric) | 76.2 mm |
Dimensions (L x W) | 3 x 3 in. (76.2 x 76.2mm) |
Color | White and Orange |
For Use With (Application) | Used for cleaning up small or large spills, to wipe pipets, probes, slide prep devices, and to pop caps from blood and specimen tubes. Wipes and Liners are non-sterile and not intended for use on patients. |
Quantity | Each |
Wipes and Liners are non-sterile and not intended for use on patients.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction