Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio-Serv™ AIN-76 Rodent Liquid Animal Diet
SDP

No blender needed—just add water, shake, and pour

$49.75

Specifications

Format Powder
For Use With (Application) Post operative recovery
Quantity 4 L
Species Rodent
View More Specs

Products 2
Catalog Number Mfr. No. Product Type Particle Shape Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Particle Shape Price Quantity & Availability  
14-726-623
SDP
Encompass
View Documents
Bio-Serv™
F1268SP
Rodent Liq. Diet CTL Shake/Bottle Powder
Each for $49.75
Only null left
Add to Cart
 
14-726-625
SDP
Encompass
View Documents
Bio-Serv™
F1268
Rodent Liquid Diet, Control Pellet
Each for $49.75
Only null left
Add to Cart
 
Description

Description

  • Easy to prepare
  • Offers the versatility of a liquid format, allowing the addition of alcohol, medications, or test compounds
  • Provides a convenient way to measure food intake or study nutritional manipulation
  • Ideal for post-operative recovery
  • Nutritionally assayed
  • Powder
  • Ingredients: Corn Syrup Solids, Casein, Corn Oil, Cellulose, Maltodextrin, Mineral Mix, Suspending Aids, Vitamin Mix, DL-Methionine, Choline Bitartrate

Specifications

Specifications

Powder
Post operative recovery
4 L
Rodent
Videos
Safety and Handling

Safety and Handling

ShelfLife : 6 Months

SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.