Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio-Serv™ Bacon Softies™ Animal Diet, Certified
SDP

Replaces labor intensive, nutrient diluted mush diets

$40.95 - $104.55

Specifications

Product Type Softies
Components Soy Flour, Soybean Oil, Corn Syrup, Sucrose, Casein, Cellulose, Corn Starch, Wheat Bran, Molasses, Mineral Mix, Banana Flakes, Acacia Gum, Bacon Flavor, Glycerin, Vitamin Mix, DL-Methionine, Choline Bitartrate, BHA
Flavor Bacon
Format Pellet
For Use With (Application) For recovering or debilitated animals and weanlings
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Sterility Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Sterility Price Quantity & Availability  
14-726-701
SDP
Encompass
View Documents
Bio-Serv™
F35801
1000 g Non-sterile
Each for $40.95
Only null left
Add to Cart
 
14-726-703
SDP
Encompass
View Documents
Bio-Serv™
F3580
5000 g Non-sterile
Each for $104.55
Only null left
Add to Cart
 
14-726-702
SDP
Encompass
View Documents
Bio-Serv™
S35801
1000 g Sterile
Each for $50.05
Only null left
Add to Cart
 
Description

Description

  • Soft, pelleted, nutritionally rich diet for rodents and supplement for canines
  • Certified (contaminant screened)
  • Nutritionally assayed

Ingredients: Soy Flour, Soybean Oil, Corn Syrup, Sucrose, Casein, Cellulose, Corn Starch, Wheat Bran, Molasses, Mineral Mix, Banana Flakes, Acacia Gum, Bacon Flavor, Glycerin, Vitamin Mix, DL-Methionine, Choline Bitartrate, BHA

Specifications

Specifications

Softies
Bacon
For recovering or debilitated animals and weanlings
Rodent, Canine
1.27 cm
Soy Flour, Soybean Oil, Corn Syrup, Sucrose, Casein, Cellulose, Corn Starch, Wheat Bran, Molasses, Mineral Mix, Banana Flakes, Acacia Gum, Bacon Flavor, Glycerin, Vitamin Mix, DL-Methionine, Choline Bitartrate, BHA
Pellet
Soft Pellet
0.5 in.
Videos
Safety and Handling

Safety and Handling

ShelfLife : 6 Months

SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.