Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio-Serv™ Bacon Yummies™ Enrichment Treats, Certified
SDP

Catalog No. 14726579
Encompass
Change view
Click to view available options
Quantity:
20,000/Bag
5000/Bg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-726-579 5000/Bg
14-726-580 20,000/Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-726-579 Supplier Bio-Serv™ Supplier No. F057781
Only null left
Add to Cart
Add to Cart

Nutritionally complete grain-based treat with a highly palatable bacon flavor

  • Excellent for reward or foraging
  • Can be fed as a sole source diet for animals with poor appetites
  • 190mg size eliminates guesswork when measuring food intake
  • Nutritionally assayed
  • Certified (Contaminant Screened)
  • Ingredients: Ground Corn, Banana Flakes, Dehulled Soybean Meal, Corn Gluten Meal, Ground Wheat, Corn Gluten Feed, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Sucrose, Fructose, Dextrose, Dried Beet Pulp, Soybean Oil, Porcine Animal Fat (preserved with BHA), Dried Brewers Yeast, Mineral Mix, Vitamin Mix, Bacon Flavor, Magnesium Stearate, Calcium Silicate, DL-Methionine, Choline Chloride, Sodium Propionate, Ascorbic Acid, Kaolin

Specifications

Product Type Bacon Yummies
Calories 3.37
Components Ground Corn, Banana Flakes, Dehulled Soybean Meal, Corn Gluten Meal, Ground Wheat, Corn Gluten Feed, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Sucrose, Fructose, Dextrose, Dried Beet Pulp, Soybean Oil, Porcine Animal Fat (preserved with BHA), Dried Brewers Yeast, Mineral Mix, Vitamin Mix, Bacon Flavor, Magnesium Stearate, Calcium Silicate, DL-Methionine, Choline Chloride, Sodium Propionate, Ascorbic Acid, Kaolin
Flavor Bacon
Format Pellet
Particle Shape Pellet
Quantity 5000/Bg
Storage Requirements Cool dry place
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.