Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio-Serv™ Fruit Crunchies™ Animal Diet, Certified
SDP

Catalog No. 14726772
Encompass
Click to view available options
Quantity:
1000 g
Sterility:
Non-sterile
Sterile

A nutritional treat with a medley of flavors

  • Nutritionally complete 190mg treat that compliments enrichment programs
  • A medley of apple, grape and orange flavors add a variety to foraging activity
  • Less labor intensive and more sanitary than perishable fruit
  • Excellent reward or foraging treat
  • Certified (Contaminant Screened)
  • Excellent source of vitamins
  • Nutritionally assayed
  • Weight: 19mg
  • Shape: pellet
  • S05798-1 is sterilized by gamma irradiation

Certifications

Contaminant screened

Specifications

Product Type Fruit Crunchies
Calories 0.66
Components Casein, Dextrose, Sucrose, Cellulose, Corn Starch, Corn Oil, Corn Syrup, Mineral Mix, Choline Bitartrate, Vitamin Mix, Natural and Artificial Flavors, Calcium Silicate, Magnesium Stearate, L-Cystine, Ascorbic Acid, Food Dye
Flavor Fruit Crunchies
Format Pellet
Packaging Quantity 1 kg/bag
Particle Shape Pellet
Quantity 1000 g
Sterility Non-sterile
Storage Requirements Cool dry place
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.