Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio-Serv™ Lieber-DeCarli '82 Rodent Liquid Diet
SDP

Catalog No. p-4185838
Encompass
Click to view available options
Product Type:
Rodent Liquid Diet, Control
Rodent Liquid Diet, Ethanol

Requires

Blender required for mixing.

Versatile liquid format allows the addition of alcohol, medications, or test compounds

  • Provides a convenient way to measure food intake or study nutritional manipulation
  • Ideal for post-operative recovery
  • Nutritionally assayed
  • Powder
  • Ingredients: Corn Syrup Solids, Casein, Olive Oil, Maltodextrin, Cellulose, Mineral Mix, Corn Oil, Suspending Aid, Safflower Oil, Vitamin Mix, L-Cystine, Choline Bitartrate, DL-Methionine

Specifications

Product Type Rodent Liquid Diet, Control
Calories 1000
Components Corn syrup solids, Casein, Olive Oil, Maltodextrin, Cellulose, Mineral Mix, Corn Oil, Suspending aids, Safflower Oil, Vitamin Mix, L-Cystine, Choline Bitartrate, DL-Methionine
Format Powder
For Use With (Application) Ideal for post operative recovery
Particle Shape Powder
Quantity 4 L
Species Rodent
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.