Learn More
Bio-Serv™ Lieber-DeCarli '82 Rodent Liquid Diet
Versatile liquid format allows the addition of alcohol, medications, or test compounds
Supplier: Bio-Serv™ F1258
Requires :
Blender required for mixing.
Description
- Provides a convenient way to measure food intake or study nutritional manipulation
- Ideal for post-operative recovery
- Nutritionally assayed
- Powder
- Ingredients: Corn Syrup Solids, Casein, Olive Oil, Maltodextrin, Cellulose, Mineral Mix, Corn Oil, Suspending Aid, Safflower Oil, Vitamin Mix, L-Cystine, Choline Bitartrate, DL-Methionine
Specifications
Rodent Liquid Diet, Ethanol | |
Corn syrup solids, Casein, Olive Oil, Maltodextrin, Cellulose, Mineral Mix, Corn Oil, Suspending aids, Safflower Oil, Vitamin Mix, L-Cystine, Choline Bitartrate, DL-Methionine | |
Ideal for post operative recovery | |
4 L |
1000 | |
Powder | |
Powder | |
Rodent |
Safety and Handling
ShelfLife : 6 Months
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.