Bio-Serv™ Mouse Diet, High Fat Paste Available on GSA/VA Contract for Federal Government customers only.

For induction and/or maintenance of obese models

Manufacturer: BIO SERV S1850

 View more versions of this product

Catalog No. 14-726-605

  Check Availability
Add to Cart

    Specifications

    Storage Requirements 6 months
    For Use With (Application) Induction and or maintenance of obese models
    Sterile Yes
    Quantity 10000g
    Species Mouse
    Components Lard, Casein, Maltodextrin, Sucrose, Mineral Mix, Vitamin Mix, DL-Methionine, Choline Chloride
    Calories 5.51
    Product Type Mouse diet
    Shelf Life 6 Months
    Particle Shape Paste

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit