Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bio-Serv™ Mouse Diet, High Fat, Soft Pellets


Click to view available options
Product Type:
Mouse Diet
Sterility:
Non-sterile
Sterile
Description
- Nutritionally complete purified diet formula successfully used in diabetes and obesity studies
- 60% kcals from fat
- Soft pellets are 0.5 in. dia. (1.3cm) and vary in length
- Nutritionally assayed
- Ingredients: Lard, Casein, Maltodextrin, Sucrose, Mineral Mix, Vitamin Mix, DL-Methionine, Choline Chloride
Specifications
Specifications
Product Type | Mouse Diet |
Calories | 5.49 |
Components | Lard, Casein, Maltodextrin, Sucrose, Mineral Mix, Vitamin Mix, DL-Methionine, Choline Chloride |
Format | Pellet |
For Use With (Application) | Nutritionally complete purified diet formula successfully used in diabetes and obesity studies |
Particle Shape | Soft Pellet |
Quantity | 5000 g |
Species | Mouse |
Sterility | Sterile |
Diameter (English) | 0.5 in. |
Show More |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction