Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bio-Serv™ PRANG™, Oral Rehydrator Powder

Click to view available options
Quantity:
100 g
1000 g
Flavor:
Apple
Banana
Cherry
Grape
Orange
Strawberry
Description
- Flavored oral rehydration with electrolytes and Vitamin C
- Balanced mineral and glucose levels for quick energy
- Provides the benefit of electrolyte drinks with less sugar
- Powder form, just add water
- Nutritionally assayed
- Certified (contaminant screened)
Ingredients: Dextrose, Sodium Chloride, Sodium Bicarbonate, Citric Acid, Potassium Chloride, Ascorbic Acid, Acesulfame Potassium, Natural and Artificial Flavors, Food Dye
Specifications
Specifications
| Product Type | Oral Rehydrator |
| Calories | 19.88 |
| Components | Electrolyte, Vitamin C, Mineral, Dextrose, Sodium Chloride, Sodium Bicarbonate, Citric Acid, Potassium Chloride, Ascorbic Acid, Acesulfame Potassium, Natural and Artificial Flavors, Food Dye |
| Flavor | Apple |
| Format | Powder |
| For Use With (Application) | Crashing, recovering, debilitating |
| Particle Shape | Powder |
| Quantity | 100 g |
| Storage Requirements | Cool dry place |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction