Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio-Serv™ PRIMA-Burger™ Grain-Based Supplement Powder
SDP

Catalog No. 14726444
Click to view available options
Quantity:
10,000 g

Adds variety to enrichment programs

  • Flavorful blend of peanut butter and molasses
  • Use frozen or as paste
  • Ideal diet to deliver and mask medications
  • Nutrionally complete
  • Nutritionally assayed

Ingredients: Ground Corn, Soybean Meal, Corn Gluten Meal, Ground Wheat, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Fructose, Molasses, Beet Pulp, Soybean Oil, Corn Oil, Porcine Animal Fat (preserved with BHA), Peanut Butter, Mineral Mix, Vitamin Mix, Ascorbic Acid

Specifications

Product Type Burger
Calories 4.03
Components Ground Corn, Soybean Meal, Corn Gluten Meal, Ground Wheat, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Fructose, Molasses, Beet Pulp, Soybean Oil, Corn Oil, Porcine Animal Fat (preserved with BHA), Peanut Butter, Mineral Mix, Vitamin Mix, Ascorbic Acid
Flavor Peanut Butter and Molasses Blend
For Use With (Application) Ideal diet to deliver and mask medications
Particle Shape Powder
Quantity 10,000 g
Storage Requirements 6 Months, Refrigerated
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.