Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BioAcryl-P (40%, 29:1), PROMO, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
Description
General Description
- BioAcryl-P is a 29:1 ratio of Acrylamide/Bisacrylamide
- Acrylamide is a crystalline amide that forms polymer structures at room temperature
- Acrylamide acts as the monomer for the gel structure, and bisacrylamide as the crosslinker for gel matrix formation
Application
- Suitable for gel electrophoresis-mediated separation of proteins and DNA for identification or sequencing

Specifications
Specifications
Chemical Name or Material | Acrylamide/Bisacrylamide |
Physical Form | Liquid |
Concentration | 40% |
Color | Colorless |
Quantity | 1 L |
MDL Number | MFCD00080848 |
UN Number | UN3426 |
Solubility Information | It is soluble in water. |
RUO - Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction