Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BioAcryl-P (40%, 39:1)

Catalog No. p-7042675
Click to view available options
Quantity:
500 mL
1 L

Acrylamide is used for sequencing gels and separation of proteins, used in protein and nucleic acid electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acrylamide is used for sequencing gels and separation of proteins, used in protein and nucleic acid electrophoresis.

Solubility
Fully miscible in water.

Notes
Protect from heat. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acrylamide/Bisacrylamide 39:1
Physical Form Liquid
Concentration 40% soln
Color Colorless
Quantity 500 mL
UN Number UN3426
Solubility Information Fully miscible in water.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.