Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BioBasic™ SCX HPLC Columns

Catalog No. 03050883
Change view
Click to view available options
Length (Metric):
10 mm
50 mm
100 mm
150 mm
250 mm
Diameter (Metric):
1 mm
2.1 mm
4 mm
4.6 mm
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
03-050-883 10 mm 4 mm
03-050-885 100 mm 2.1 mm
03-050-887 100 mm 4.6 mm
03-050-888 150 mm 1 mm
03-050-890 250 mm 1 mm
73205254630 250 mm 4.6 mm
03-050-884 50 mm 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 03-050-883 Supplier Thermo Scientific™ Supplier No. 73205014001
Add to Cart
Add to Cart

BioBasic SCX LC Columns are optimized for the separation of proteins, peptides and other cationic species.

Use Thermo Scientific™ BioBasic™ SCX LC Columns for the optimized separation of proteins, peptides and other cationic species. These high-performance columns are ideal for the analysis of proteins, peptides and other biomolecules with higher efficiency than polymeric ion exchange.

  • Strong cation exchange phase based on sulfonic acid chemistry
  • Separation and retention of basic and other cationic species
  • 300Å pore size for enhanced protein and peptide separations
  • Outstanding stability under demanding pH conditions

Ordering Information:

Other column dimensions are available. Please call Customer Service for more information.

TRUSTED_SUSTAINABILITY

Specifications

Column Format Guard Cartridge, 4 Pk.
Diameter (Metric) 4 mm
Length (Metric) 10 mm
Max. Pressure 5800 psi (400 bar)
Particle Size 5 μm
pH 2 to 8
Pore Size 300 Å
Surface Area 100 m²/g
Temperature 60°C
USP Type L52
Stationary Phase Sulfonic Acid
Product Line BioBasic
Packing Material Silica
Carbon Load 3%
Column Type Ion Exchange
Endcapped No
Phase Ion Exchange
Technique Bio-LC
Product Type Guard Cartridge
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.