Learn More
Thermo Scientific™ BioBasic™ SEC 1000 Size Exclusion Chromatography HPLC Columns
Shop All Thermo Scientific ProductsDescription
Offer outstanding size separation of water soluble compounds with BioBasic™ SEC 1000 Size Exclusion Chromatography HPLC Columns.
Offer outstanding size separation of water soluble compounds with Thermo Scientific™ BioBasic™ SEC 1000 Size Exclusion HPLC Columns. BioBasic SEC columns provide high efficiency separations for a wide range of molecular weight from 100 to 10,000,000 daltons allowing the analysis of proteins, peptides and other biomolecules.
- Covers separation of analytes over a wide molecular weight range
- Long column life and high column efficiencies
- Simple mechanism of interaction based on molecular size and shape
- Ideal for sample cleanup
- Straightforward method development, simple mobile phases
- 1000Å pore size
Ordering Information:
For information on bulk quantities, please inquire.
Specifications
Specifications
| Column Format | High Temperature Column |
| Diameter (Metric) | 7.8 mm |
| Length (Metric) | 300 mm |
| Max. Pressure | 5800 psi (400 bar) |
| Particle Size | 5 μm |
| Pore Size | 1000 Å |
| USP Type | L33 |
| Stationary Phase | Diol (OH) |
| Product Line | BioBasic |
| Packing Material | Spherical, Fully Porous, Ultrapure Silica |
| Show More |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.