Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BioBasic™ SEC 120 Size Exclusion Chromatography HPLC Columns

Catalog No. 03050894 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
300 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
03-050-894 300 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-050-894 Supplier Thermo Scientific™ Supplier No. 73405307846

missing translation for 'validMainframeMsg'

Outstanding size separation of water soluble compounds.

Provide outstanding size separation of water soluble compounds with Thermo Scientific™ BioBasic™ SEC 120 Size Exclusion Chromatography HPLC Columns. BioBasic SEC columns provide high efficiency separations for a wide range of molecular weights from 100 to 10,000,000 daltons, allowing the analysis of proteins, peptides and other biomolecules.

  • Covers separation of analytes over a wide molecular weight range
  • Long column life and high column efficiencies
  • Simple mechanism of interaction based on molecular size and shape
  • Excellent for sample cleanup
  • Straightforward method development, simple mobile phases
  • 120Å pore size

Ordering Information:

For information on bulk quantities, please inquire.

TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column
Diameter (Metric) 7.8 mm
For Use With Size Exclusion Chromatography
Length (Metric) 300 mm
Max. Pressure 5800 psi (400 bar)
Particle Size 5 μm
Pore Size 120 Å
USP Type L33
Stationary Phase Diol (OH)
Product Line BioBasic
Packing Material Spherical, Fully Porous, Ultrapure Silica
Column Type Size Exclusion
Phase Size Exclusion
Product Type Analytical Column
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.