Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BioBasic™ 8 5μm Nanobore HPLC Columns

Catalog No. 03170367 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
50 mm
100 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
03-170-367 50 mm
03-251-539 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-170-367 Supplier Thermo Scientific™ Supplier No. 72205057581

missing translation for 'validMainframeMsg'

A greater signal to noise ratio than traditional columns used with electrospray at flowrates of nL/min. versus mL/min.

At flow rates of nL/min versus mL/min, Thermo Scientific™ BioBasic 8 5μm Nanobore HPLC Columns offer greater signal to noise ratio than traditional columns used with electrospray.

  • High sensitivity flexible silica capillary columns
  • Optimized for LC/MS applications

Features of BioBasic 8 Solid Phase

  • 300Å pore size for improved biomolecule analysis
  • An excellent starting column for protein and peptide analysis
  • Outstanding reproducibility, efficiency and column lifetime
  • Optimized for LC/MS separations

Ordering Information:

Unless otherwise specified, IntegraFrit and PicoFrit are sold in single-column units.

TRUSTED_SUSTAINABILITY

Specifications

Column Format Nanobore Column
Diameter (Metric) 75 μm
For Use With Nanospray LC/MS
Length (Metric) 50 mm
Particle Size 5 μm
pH 2 to 10
Pore Size 300 Å
USP Type L7
Stationary Phase C8
Packing Material Silica
Carbon Load 5%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.