Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BioBasic™ 8 HPLC Columns

Catalog No. p-4523550 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
10 mm
250 mm
Diameter (Metric):
4 mm
4.6 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
03-050-868 250 mm 4.6 mm
03-051-671 10 mm 4 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-050-868 Supplier Thermo Scientific™ Supplier No. 72205254630
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use this column for reversed-phase separation of proteins and other biomolecules.

Optimized for the separation of a wide range of peptides, Thermo Scientific™ BioBasic™ 8 LC Columns are high-performance columns ideal for the analysis of proteins, peptides and other biomolecules.

  • 300Å pore size for improved biomolecule analysis
  • An excellent starting column for protein and peptide analysis
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations

Ordering Information:

Other column dimensions are available, including preparative columns. Please call Customer Service for more information.

TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column
Diameter (Metric) 4.6 mm
Length (Metric) 250 mm
Max. Pressure 5800 psi (400 bar)
Particle Size 5 μm
pH 2 to 9
Pore Size 300 Å
Surface Area 100 m²/g
Temperature 60°C
USP Type L7
Stationary Phase C8
Product Line BioBasic
Packing Material Spherical, Fully Porous, Ultrapure Silica
Carbon Load 5%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Technique Bio-LC
Product Type Analytical Column
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.