Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BioBasic™ 8 PEEK Bio-Inert HPLC Columns, 5μm Particle Size

Catalog No. 72205102168
Change view
Click to view available options
Length (Metric):
100 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
72205102168 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 72205102168 Supplier Thermo Scientific™ Supplier No. 72205102168

Thermo Scientific™ BioBasic 8 PEEK Bio-Inert HPLC Columns, 5μm Particle Size, are optimized for the separation of a wide range of peptides.

Thermo Scientific™ BioBasic 8 PEEK Bio-Inert HPLC Columns, 5μm Particle Size, are optimized for the separation of a wide range of peptides. These high-performance columns are ideal for the analysis of proteins, peptides and other biomolecules. Optimized for the separation of medium hydrophobicity peptides, nominally less than 20,000 daltons.

  • 300Å pore size for improved biomolecule analysis
  • An excellent starting column for protein and peptide analysis
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations

Ordering Information:

Other column dimensions are available in bio-inert column hardware. Please call Customer Service for more information.

TRUSTED_SUSTAINABILITY

Specifications

Column Format PEEK Column
Diameter (Metric) 2.1 mm
For Use With LC/MS
Length (Metric) 100 mm
Particle Size 5 μm
pH 2 to 10
Pore Size 300 Å
USP Type L7
Stationary Phase C8
Packing Material Silica
Carbon Load 5%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.