Learn More
Description
- Covers separation of analytes over a wide molecular weight range
- Long column life and high column efficiencies
- Simple mechanism of interaction based on molecular size and shape
- Excellent for sample cleanup
- Straghtforward method development, simple mobile phases
- 300Å pore size
Order Info
For information on bulk quantities, please inquire.
Specifications
Specifications
| Diameter (Metric) | 1 mm |
| Length (Metric) | 250 mm |
| Particle Size | 5 μm |
| Pore Size | 300 Å |
| USP Type | L33 |
| Stationary Phase | Proprietary |
| Packing Material | Silica |
| Column Type | Size Exclusion |
| Phase | Size Exclusion |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.