Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BioBasic™ SEC 300A HPLC Columns

Catalog No. p-3136988 Shop All Thermo Scientific Products
Change view
Click to view available options
Diameter (Metric):
1 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
73505251030 1 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 73505251030 Supplier Thermo Scientific™ Supplier No. 73505251030

missing translation for 'validMainframeMsg'

Outstanding size separation of water soluble compounds

  • Covers separation of analytes over a wide molecular weight range
  • Long column life and high column efficiencies
  • Simple mechanism of interaction based on molecular size and shape
  • Excellent for sample cleanup
  • Straghtforward method development, simple mobile phases
  • 300Å pore size

Order Info

For information on bulk quantities, please inquire.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 1 mm
Length (Metric) 250 mm
Particle Size 5 μm
Pore Size 300 Å
USP Type L33
Stationary Phase Proprietary
Packing Material Silica
Column Type Size Exclusion
Phase Size Exclusion
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.