Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Biochemistry Kits
SCI_ED

Catalog No. s30817
Click to view available options
Product Type:
Biochemistry II

Kemtec™ Biochemistry Kits provide teachers with the resources to develop course material, and experiments at varying complexities

Biochemistry I:

  • Introduce students to the chemistry of living things
  • Features five classic biochemistry experiments
  • Use Benedicts reagent, Biuret, Sudan IV, Iodine to test for proteins, carbohydrates, and lipids
  • Use pepsin to digest protein
  • Test the activity of salivary amylase (both in vitro and in vivo)
  • Develop a technique for determining ascorbic acid concentration to use to evaluate Vitamin C levels
  • Determine of the alkalinity of acidity of foods
  • Ideal for students in biology, general science, nutritional science, or biochemistry

Biochemistry II:

  • Includes an entire course worth of classic biochemistry experiments presented at a more in-depth level than Biochemistry I
  • Learn how to measure the pH of foods, saliva, and soil
  • Learn classic detection methods for sugars, starch, fat, and protein
  • Pepsin digestion of protein
  • Amino acid detection using ninhydrin
  • Explore the effects of varying conditions on salivary amylase activity
  • Model how digestion works
  • Determine ascorbic acid concentration
  • Explore dyes and plant pigments using chromatography with different solvents
  • Ideal for supporting higher level biology, nutritional science, advanced general science, or biochemistry courses

Specifications

Product Type Biochemistry II
Grade 10 to 14
Age 13 to 17
For Use With (Application) Grades 10-14
Includes Instructor's Manual, Chromatography Vials/Solvent Solutions, Dialysis Tubing, Benedict's Solution, Biret Solution, Hydrochloric Acid, Sodium Hydroxide, Starch, Glucose, Albumin, Pepsin, Ascorbic Acid, Iodine Solution, Sudan IV Solution, Sodium Bicarbonate, Acetone, Vegetable Oil, Ninhydrin, 2,6-Dichloroindophenol, Dye, Amino Acid Mixture, pH Paper, Pipets
Quantity 1 Kit
Requires Hotplate, 50 mL Beakers, Forceps, Storage Jars/Caps, Funnels, Food Samples (Not Included)
Time Range 8 to 10 hr.
Weight (English) 4 lb.
Dimensions (L x W x H) 13 x 12 x 5 in.
Class Size 24 Students
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.