Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Biohazard Clamp On Shield
SDP

Catalog No. p-7078153
Click to view available options
Width (English):
12 in.
Length (English):
8.5 in.
12 in.
This item is not returnable. View return policy
This item is not returnable. View return policy

Protect user from splash and biohazard exposure. Poltex Clamp On Biohazard Shield is made from polycarbonate.

  • Mounts to virtually any surface with a powerful clamp
  • Made from abrasion and break- resistant polycarbonate
  • Flexible neck for maximum adjustability
  • Available in two size: 8.5 x 12 in. and 12 x 12 in.

Specifications

Dimensions (L x W) 8.5 x 12 in.
Width (Metric) 30.5 cm
Width (English) 12 in.
Product Type Biohazard Clamp-on Shield
Length (English) 8.5 in.
Material Polycarbonate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.