Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Biohazard Labels Vinyl BIOHAZARD
Shop All Brady Corporation Products
Click to view available options
Dimensions (LxW):
10 x 7 in.
108 ft. x 2 in.
2.875 x 2 in.
3.5 x 5 in.
5 x 3.5 in.
Description
- Good resistance to most common chemicals, oils and water
- Warning labels support compliance with OSHA 29 CFR 1910.1030 Regulation for Bloodborne Pathogens Labeling
- Warning labels support compliance with DOT 49 CFR 172.400(b) Labeling Requirements for transportation
- Service temperature of -40°F to +122°F (-40°C to +50°C)
Specifications
Specifications
| Material | Vinyl |
| No. per Pack | 10/Pk. |
| Dimensions (LxW) | 10 x 7 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction