Learn More
Therapak™ Biological Substance Category B Replenishment Supplies
Catalog No. 22130060 Shop All Therapak Corporation Products
Change view
Click to view available options
Includes:
3 oz. Refrigerant Pack
8 oz. Refrigerant Pack
Large Insulated EPS Cooler
Medium Insulated EPS Cooler
Small Insulated EPS Cooler
Size:
3 oz. pack
8 oz. pack
Large
Medium
Small
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Size
22-130-060 Large Insulated EPS Cooler Large
22-130-059 Medium Insulated EPS Cooler Medium
22-130-072 8 oz. Refrigerant Pack 8 oz. pack
22-130-414 3 oz. Refrigerant Pack 3 oz. pack
22-130-058 Small Insulated EPS Cooler Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 22-130-060 Supplier Therapak™ Supplier No. 37910
Add to Cart
Add to Cart
For shipment of biological specimens
  • Insulated Shippers include standard DOT/IATA labels, EPS cooler, and corrugated shipper box
  • Leakproof Refrigerant Shippers keep lab specimens cold (30°F) with nontoxic, food-grade gel refrigerants
Specifications
Includes Large Insulated EPS Cooler
Applications For shipment of Biological Substance Category B specimens
Size Large
Dimensions (LxWxH) Interior: 16 x 13 x 11 in.Exterior: 18 x 16 x 14 in.