Learn More
Vernier ProScope HR Digital Microscope Kits
Shop All Vernier Software Products
Description
The ProScope HR is a USB handheld digital microscope designed for use with computer. With a high-quality CMOS sensor and universal lens mount, the ProScope HR is a powerful addition to any classroom.
Biology Kit:
Use ProScope HR as dissecting microscope or attach ProScope HR to regular microscope for even more magnification.
Includes:
- ProScope HR base
- ProScope HR software
- 50X lens
- 1-10X lens
- C-mount adapter with IR cut filter
- Microscope lens tube adapter
- Camera stand
- Rugged hard-side case
Deluxe Kit:
For maximum magnification with or without regular microscope, deluxe kit has covered with its lens ensemble and accessories.
Includes:
- ProScope HR base
- ProScope HR software
- 50X lens
- 1-10X lens
- 100X lens
- 200X lens
- C-mount adapter with IR cut filter
- Microscope lens tube adapter
- Camera stand
- Rugged hard-side case
Proscope HR Specifications:
- Image sensor: 1/4 in. high resolution 1.3 megapixel sensor
- Connection type: USB
- Image capture resolutions:
- 320x240 pixels
- 640x480 pixels
- 1280x1024 pixels
- Video capture resolutions:
- 320x240 pixels at @30 fps
- 640x480 pixels at @15 fps
- 1280x1024 pixels at @3.75 fps
Specifications
Specifications
Description | Biology ProScope HR Kit |
Includes | ProScope HR base and software, 50X lens, 1-10X lens, C-mount adapter with IR cut filter, microscope lens tube adapter, camera stand and rugged hard-side case |
Required Accessories | Windows: Windows™ XP service pack 2 or newer, Quicktime 7.0 or newer, DirectX 9 or newer, available USB 2 port, Pentium 4 or equivalent; Macintosh: Mac OS 10.4.8 or newer, Quicktime 7.0 or newer, factory-installed USB 2 port, G4, G5, or Intel processor; Chromebook: Camera app and available USB 2 port |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.