Description
Includes
5 x 0.01g Adenosine 5’-Triphosphate, Disodium Salt and 5 x 0.017g Firefly Lantern Extract Powder
Specifications
Specifications
| Product Type | Bioluminescence Chemistry Demo Kit |
| For Use With (Application) | Study the concept of bioluminescence using real firefly extract. |
| Includes | 5 x 0.01g Adenosine 5’-Triphosphate, Disodium Salt and 5 x 0.017g Firefly Lantern Extract Powder |