Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Biomedical Polymers Translucent Multipurpose Containers
Click to view available options
Capacity (Metric):
237 mL
473 mL
946 mL
1900 mL
2500 mL
5100 mL
Description
- Ideal for storing biologically hazardous samples for terminal autoclaving before disposal
- Molded from either high-density polyethylene or polypropylene, containers are inert to most chemicals
- Resistant to freezing and boiling
- Easy to write on with either pen or pencil
Specifications
Specifications
Capacity | 64 oz., 1900 mL |
Capacity (Metric) | 1900 mL |
Capacity (English) | 64 oz. |
Color | Translucent |
Material | Polyethylene |
Closure Color | Translucent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction