Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab™ BioMesh Biopsy Bags
SDP

Catalog No. p-7227793
Click to view available options
Dimensions (L x W):
30 x 45 mm
45 x 75 mm
This item is not returnable. View return policy
This item is not returnable. View return policy

Synthetic mesh and small pore design ensures unrestricted flow of processing reagents around specimens.

  • Bags secure small biopsies during processing
  • Resistant to solvents and acids

Specifications

Product Type Biopsy Bag
Material Polyester
Color White
Length (Metric) 45 mm
Width (Metric) 30 mm
Dimensions (L x W) 30 x 45 mm
Size Small
Quantity 1000/Cs.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.