Learn More
StatLab BioMesh Cassettes
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Pink
Red
Tan
Yellow
Description
- Lids separate, ideal for small, fragile specimens
- Excellent for use with Tissue Guard processing gel
- Keep small biopsies and fragile specimens secure with these fine mesh cassettes that eliminate the need for messy sponges, bags and paper
- Rounded BioMesh edges ensure no tissue fragments are lost in the corners
Specifications
Specifications
Quantity | 250 Pk. |
Color | Pink |