Learn More
StatLab BioMesh Cassettes
SDP
Catalog No. p-7228696
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Pink
Red
Tan
Yellow
This item is not returnable. View return policy
This item is not returnable. View return policy
StatLab BioMesh Cassettes keep small biopsy samples secure and are ideal for automated cassette printers.
  • Lids separate, ideal for small, fragile specimens
  • Excellent for use with Tissue Guard processing gel
  • Keep small biopsies and fragile specimens secure with these fine mesh cassettes that eliminate the need for messy sponges, bags and paper
  • Rounded BioMesh edges ensure no tissue fragments are lost in the corners
Specifications
Quantity 250 Pk.
Color Blue
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.