Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LaMotte™ BioPaddles™ Dual-Agar Paddles for Environmental Education

Shop All LaMotte Company Products


Click to view available options
For Use With (Application):
Bacterial culture
Bacterial isolation, testing
Fungi, yeast, mold isolation
Microbial culture, enumeration
Description
Includes
Free basic microbe identification app and access to technical information sheet
Flexible, ready-to-use paddles identify and quantify microbes in air, soil, water or on any surface.
- Longer shelf life than traditional petri dishes
- No refrigeration required
- No need for inoculating loops or Bunsen burners
- Requires simple incubation and a magnifier
- Unique design enhances colony morphology characterization
- Ten paddles per box
- Nutrient Agar: For routine culture of non-fastidious bacteria
- Sabouraud Dextrose Agar: For selective cultivation of fungi (yeasts and molds)
- Tryptic Soy (TSA)/Rose Bengal (RB) Agar: For cultivation of a wide variety of microorganisms (TSA) and selective isolation of yeasts and molds (RB)
- Nutrient TTC/MacConkey: For field sampling cultivation and enumeration of coliform bacteria total coliform count, Gram (-) bacterial colonies appear as red dots, Gram (+) bacteria are usually inhibited. MAC: Medium gives improved differentiation between coliforms and nonlactose fermenting organisms; Gram (+) cocci are usually inhibited
Order Info
Ships on Monday and Tuesday ONLY.
Specifications
Specifications
Content And Storage | 18°C to 25°C |
Manufacturing Origin | United States |
Media Type | TSA/RB dual Agar |
Sterility | Sterile |
For Use With (Application) | Fungi, yeast, mold isolation |
Includes | Free basic microbe identification application and access to technical information sheet |
Packaging Type | Paddle |
Quantity | 40 mL |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction