Learn More
Description
- The particles are functionalized with a proprietary polyphenyl bonding process that was specifically designed for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments.
Specifications
Specifications
| Diameter (Metric) | 4.6 mm |
| Length (Metric) | 150 mm |
| Particle Size | 2.7 μm |
| Pore Size | 450 Å |
| Quantity | 1/Ea. |
| Product Type | Column |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.