Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ BIOshell™ A160 Peptide C18 Column, 2 μm Particle Size

Catalog No. 111006759 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
2 cm
3 cm
5 cm
7.5 cm
10 cm
15 cm
Diameter (Metric):
2.1 mm
3 mm
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
11-100-6759 15 cm 3 mm
11-100-7247 5 cm 2.1 mm
11-100-6981 10 cm 2.1 mm
11-100-6758 15 cm 2.1 mm
11-100-6982 10 cm 3 mm
11-100-7245 2 cm 2.1 mm
11-100-7246 3 cm 2.1 mm
11-100-7122 7.5 cm 2.1 mm
11-100-7248 2 cm 3 mm
11-100-7249 3 cm 3 mm
11-100-7250 5 cm 3 mm
11-100-7123 7.5 cm 3 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 11-100-6759 Supplier MilliporeSigma™ Supelco™ Supplier No. 67277U
Only null left
Add to Cart
Add to Cart

Shell Thickness: 0.4 μm. Solid Core: 1.2 μm. Extent of labeling: 4.0%

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 3 mm
Length (Metric) 15 cm
Particle Size 2 μm
Pore Size 160 Å
Surface Area 65 m2/g
Stationary Phase C18 (octadecyl)
Product Line BIOshell
Matrix C18 (octadecyl) Phase/Spherical Silica Particle Platform
Endcapped No
Phase Reversed Phase
Material Stainless Steel Hardware
Technique Fused-Core HPLC
Show More Show Less

BIOshell is a trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.