Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ BIOshell™ A160 Peptide C18 Column, 2 μm Particle Size

Catalog No. 111007247 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
2 cm
3 cm
5 cm
7.5 cm
10 cm
15 cm
Diameter (Metric):
2.1 mm
3 mm
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
11-100-7247 5 cm 2.1 mm
11-100-6981 10 cm 2.1 mm
11-100-6758 15 cm 2.1 mm
11-100-6982 10 cm 3 mm
11-100-6759 15 cm 3 mm
11-100-7245 2 cm 2.1 mm
11-100-7246 3 cm 2.1 mm
11-100-7122 7.5 cm 2.1 mm
11-100-7248 2 cm 3 mm
11-100-7249 3 cm 3 mm
11-100-7250 5 cm 3 mm
11-100-7123 7.5 cm 3 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 11-100-7247 Supplier MilliporeSigma™ Supelco™ Supplier No. 67239U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Shell Thickness: 0.4 μm. Solid Core: 1.2 μm. Extent of labeling: 4.0%

  • Stationary Phase: C18 (octadecyl)
  • Particle Platform: Superficially porous (Fused-Core®)
  • Pore Size: 160 Å
  • Surface Area: 65 m2/g
  • Operating pH Range: 1 to 8
  • Maximum Pressure: 14,500 psi (1000 bar)
  • Maximum Temperature: 60°C
  • USP Classification: L1
  • Suitability: HPLC, UHPLC, LC/MS, and UHPLC-MS
  • Application: Biomolecule Separation
  • Shell Thickness: 0.4 μm
  • Solid Core: 1.2 μm
  • Extent of Labeling: 4.0%
  • Endcapped: No
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 2.1 mm
Length (Metric) 5 cm
Particle Size 2 μm
Pore Size 160 Å
Surface Area 65 m2/g
Stationary Phase C18 (octadecyl)
Product Line BIOshell
Matrix C18 (octadecyl) Phase/Spherical Silica Particle Platform
Endcapped No
Phase Reversed Phase
Material Stainless Steel Hardware
Technique Fused-Core HPLC
Show More Show Less

BIOshell is a trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.