Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ BIOshell™ A160 Peptide C18 HPLC Column, 160 Å, 4.7 μm Particle Size

Catalog No. 111008369 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
3 cm
5 cm
7.5 cm
10 cm
15 cm
Diameter (Metric):
2.1 mm
3 mm
4.6 mm
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
11-100-8369 3 cm 2.1 mm
11-100-7926 5 cm 2.1 mm
11-100-7512 10 cm 2.1 mm
11-100-7057 15 cm 2.1 mm
11-100-7927 5 cm 3 mm
11-100-7513 10 cm 3 mm
11-100-7058 15 cm 3 mm
11-100-7928 5 cm 4.6 mm
11-100-7514 10 cm 4.6 mm
11-100-7059 15 cm 4.6 mm
11-100-7669 7.5 cm 2.1 mm
11-100-8370 3 cm 3 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 11-100-8369 Supplier MilliporeSigma™ Supelco™ Supplier No. 67001U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BIOshell™ 160 Å Peptide C18 columns are specifically engineered to provide efficient separation of peptides as well as small proteins.

BIOshell™ 160 Å Peptide C18 columns contain advanced Fused-Core particles with pores strategically sized to 160 Å to enable optimized peptide diffusion. This attribute makes these columns an excellent choice for peptide mapping. Additionally, the sterically-protected C18 ligands provide extra stability allowing the columns to be used at an extended pH range (2-9) and high temperatures (up to 90°C). This greatly expands the application range for the separation of biomolecules
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 2.1 mm
Length (Metric) 3 cm
Particle Size 4.7 μm
pH 1 to 8
Pore Size 160 Å
Surface Area 60 m2/g
Stationary Phase C18 (octadecyl)
Product Line BIOshell
Matrix C18 (octadecyl) Phase/Fused-Core Particle Platform
Carbon Load 4%
Endcapped No
Phase Reversed Phase
Material Stainless Steel
Technique Fused-Core HPLC
Show More Show Less

BIOshell is a trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.